missing translation for 'onlineSavingsMsg'
Learn More

1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene, 99+%, Thermo Scientific™

Product Code. 11347359
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347359 10 g 10g
11337359 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347359 Supplier Thermo Scientific Alfa Aesar Supplier No. H27476.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95480-29-8
Molecular Formula C21H24O
Molecular Weight (g/mol) 292.422
MDL Number MFCD01218020
InChI Key QRMMWBMCJKXXKV-UHFFFAOYSA-N
Synonym 1-4-ethoxyphenyl ethynyl-4-n-pentylbenzene, 1-2-4-ethoxyphenyl ethynyl-4-pentylbenzene, 1-ethoxy-4-4-pentylphenyl ethynyl benzene, 1-ethoxy-4-2-4-pentylphenyl ethynyl benzene, 4-ethoxy-1-2-4-pentylphenyl ethynyl benzene, 1-4-ethoxyphenyl ethynyl-4-pentylbenzene #, benzene, 1-ethoxy-4-4-pentylphenyl ethynyl
PubChem CID 610223
IUPAC Name 1-ethoxy-4-[2-(4-pentylphenyl)ethynyl]benzene
SMILES CCCCCC1=CC=C(C=C1)C#CC2=CC=C(C=C2)OCC

Specifications

Melting Point 62°C (N)
Boiling Point 178°C to 180°C (0.5 mmHg)
Quantity 10 g
Formula Weight 292.42
Percent Purity ≥99%
Chemical Name or Material 1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.