CAS RN 95480-29-8
CAS RN 95480-29-8
IUPAC Name:
1-[2-(4-ethoxyphenyl)ethynyl]-4-pentylbenzene
Synonyms:
1-[2-(4-ethoxyphenyl)ethynyl]-4-pentylbenzene
Molecular Weight (g/mol):
292.42
Molecular Formula:
C21H24O
InChi Key:
QRMMWBMCJKXXKV-UHFFFAOYSA-N
SMILES:
CCCCCC1=CC=C(C=C1)C#CC1=CC=C(OCC)C=C1
1
–
1
of
1
results
1
1-[(4-Ethoxyphenyl)ethynyl]-4-n-pentylbenzene, 99+%, Thermo Scientific™
CAS: 95480-29-8 Molecular Formula: C21H24O Molecular Weight (g/mol): 292.422 MDL Number: MFCD01218020 InChI Key: QRMMWBMCJKXXKV-UHFFFAOYSA-N Synonym: 1-4-ethoxyphenyl ethynyl-4-n-pentylbenzene,1-2-4-ethoxyphenyl ethynyl-4-pentylbenzene,1-ethoxy-4-4-pentylphenyl ethynyl benzene,1-ethoxy-4-2-4-pentylphenyl ethynyl benzene,4-ethoxy-1-2-4-pentylphenyl ethynyl benzene,1-4-ethoxyphenyl ethynyl-4-pentylbenzene #,benzene, 1-ethoxy-4-4-pentylphenyl ethynyl PubChem CID: 610223 IUPAC Name: 1-ethoxy-4-[2-(4-pentylphenyl)ethynyl]benzene SMILES: CCCCCC1=CC=C(C=C1)C#CC2=CC=C(C=C2)OCC