Learn More
1-(4-Bromophenyl)pyrrolidin-2-one, 98%
Product Code. 11420030
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420030 5 g 5g
11410030 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11420030 Supplier Thermo Scientific Alfa Aesar Supplier No. H50563.06
 Restricted Item
In Stock
Add to basket
CAS: 7661-32-7 | C10H10BrNO | 240.1 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7661-32-7
Molecular Formula C10H10BrNO
Molecular Weight (g/mol) 240.1
MDL Number MFCD00138502
InChI Key YINFEFUSAQRZGG-UHFFFAOYSA-N
Synonym 1-4-bromophenyl pyrrolidin-2-one, 1-4-bromophenyl-2-pyrrolidinone, 2-pyrrolidinone, 1-4-bromophenyl, 1-4-bromophenyl-2-pyrrolidone, pubchem9356, p-bromo-phenylpyrrolidone, acmc-209p4u, buttpark 32\07-61, n-4-bromophenyl-2-pyrrolidinone, 1-4-bromophenyl pyrrolidine-2-one
PubChem CID 736110
IUPAC Name 1-(4-bromophenyl)pyrrolidin-2-one
SMILES C1CC(=O)N(C1)C2=CC=C(C=C2)Br
Specifications
Boiling Point 98°C to 101°C
Quantity 5 g
Formula Weight 240.1
Percent Purity 98%
Chemical Name or Material 1-(4-Bromophenyl)pyrrolidin-2-one