accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BrNO

CAS RN 7661-32-7

IUPAC Name: 1-(4-bromophenyl)pyrrolidin-2-one
Synonyms: 1-(4-bromophenyl)pyrrolidin-2-one
Molecular Weight (g/mol): 240.1
Molecular Formula: C10H10BrNO
InChi Key: YINFEFUSAQRZGG-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N1CCCC1=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results