Learn More
1-[4-(1,1-Dimethylpropyl)phenyl]-4-[4-(diphenylmethoxy)piperidin-1-yl]butan-1-one Hydrogen Fumarate, Mikromol™
Description
1-[4-(1,1-Dimethylpropyl)phenyl]-4-[4-(diphenylmethoxy)piperidin-1-yl]butan-1-one Hydrogen Fumarate
Specifications
Specifications
| Chemical Name or Material | 1-[4-(1,1-Dimethylpropyl)phenyl]-4-[4-(diphenylmethoxy)piperidin-1-yl]butan-1-one Hydrogen Fumarate |
| CAS | 2734346-74-6 |
| Analyte or Component Names | 1-[4-(1,1-Dimethylpropyl)phenyl]-4-[4-(diphenylmethoxy)piperidin-1-yl]butan-1-one Hydrogen Fumarate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C33 H41 N O2 . C4 H4 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ebastine Imp. E (EP), 1-[4-(1,1-Dimethylpropyl)phenyl]-4-[4-(diphenylmethoxy)piperidin-1-yl]butan-1-one Hydrogen Fumarate, Ebastine Imp. E (EP) as Hydrogen Fumarate, Ebastine Related Compound E as Fumarate, Ebastine Impurity E as Hydrogen Fumarate |
| InChI Formula | InChI=1S/C19H20F3NO2.ClH/c1-14(13-16-9-5-6-10-17(16)19(20,21)22)23-11-12-25-18(24)15-7-3-2-4-8-15;/h2-10,14,23H,11-13H2,1H3;1H |
| SMILES | CCC(C)(C)c1ccc(cc1)C(=O)CCCN2CCC(CC2)OC(c3ccccc3)c4ccccc4.OC(=O)\C=C\C(=O)O |
| IUPAC Name | 4-(4-benzhydryloxypiperidin-1-yl)-1-[4-(2-methylbutan-2-yl)phenyl]butan-1-one;(E)-but-2-enedioic acid |
| Show More |