Learn More
1,3-Bis(2-methoxyphenoxy)propan-2-ol, Mikromol™
Product Code. 30051209
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051209 25 mg 25mg
17913956 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30051209 Supplier Mikromol Supplier No. MM0354.030025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,3-Bis(2-methoxyphenoxy)propan-2-ol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,3-Bis(2-methoxyphenoxy)propan-2-ol
CAS 16929-60-5
Analyte or Component Names 1,3-Bis(2-methoxyphenoxy)propan-2-ol
Shipping Condition Room Temperature
Molecular Formula C17 H20 O5
Certifications/Compliance ISO 17025
Synonym Guaifenesin Imp. D (EP), 1,3-Bis(2-methoxyphenoxy)propan-2-ol, Guaifenesin Impurity D
InChI Formula InChI=1S/C17H20O5/c1-19-14-7-3-5-9-16(14)21-11-13(18)12-22-17-10-6-4-8-15(17)20-2/h3-10,13,18H,11-12H2,1-2H3
SMILES COc1ccccc1OCC(O)COc2ccccc2OC
IUPAC Name 1,3-bis(2-methoxyphenoxy)propan-2-ol
Molecular Weight (g/mol) 304.34
Quantity 25 mg
Formula Weight 304.1311 g/mol