Learn More
1-[[3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]piperazine (Desmethylsildenafil), Mikromol™
Description
1-[[3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]piperazine (Desmethylsildenafil), Mikromol
Specifications
Specifications
| Chemical Name or Material | Desmethylsildenafil (1-[(3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl)sulfonyl]piperazine) |
| CAS | 139755-82-1 |
| Analyte or Component Names | Desmethylsildenafil (1-[(3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl)sulfonyl]piperazine) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H28 N6 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-[[3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]piperazine, Desmethylsildenafil, Sildenafil Citrate Impurity F (EP), Sildenafil Impurity F (EP) |
| InChI Formula | InChI=1S/C21H28N6O4S/c1-4-6-16-18-19(26(3)25-16)21(28)24-20(23-18)15-13-14(7-8-17(15)31-5-2)32(29,30)27-11-9-22-10-12-27/h7-8,13,22H,4-6,9-12H2,1-3H3,(H,23,24,28) |
| Solution Type | Neat |
| SMILES | CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCNCC4 |
| Show More |