Learn More
1-(2-Hydroxyphenoxy)-3-(isopropylamino)-2-propanol Hydrochloride, Mikromol™
Product Code. 30052325
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052325 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30052325 Supplier Mikromol Supplier No. MM0129.010025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(2-Hydroxyphenoxy)-3-(isopropylamino)-2-propanol Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(2-Hydroxyphenoxy)-3-(isopropylamino)-2-propanol Hydrochloride
CAS 2829-92-7
Analyte or Component Names 1-(2-Hydroxyphenoxy)-3-(isopropylamino)-2-propanol Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C12 H19 N O3 . Cl H
Certifications/Compliance ISO 17025
Synonym 1-(2-Hydroxyphenoxy)-3-(isopropylamino)-2-propanol Hydrochloride
InChI Formula InChI=1S/C12H19NO3.ClH/c1-9(2)13-7-10(14)8-16-12-6-4-3-5-11(12)15;/h3-6,9-10,13-15H,7-8H2,1-2H3;1H
SMILES Cl.CC(C)NCC(O)COc1ccccc1O
IUPAC Name 2-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenol;hydrochloride
Molecular Weight (g/mol) 261.75
Quantity 25 mg
Formula Weight 261.1132 g/mol