Learn More
1,2-Diphenyl-3,5-pyrazolidinedione (>90%), TRC
Product Code. 30377990
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377990 2.5 g 2.5g
30385857 250 mg -
30383437 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30377990 Supplier Toronto Research Chemicals Supplier No. TRCD4922652.5G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2652-77-9
Molecular Formula C15H12N2O2
Molecular Weight (g/mol) 252.27
Synonym 1,2-Diphenyl-3,5-dioxopyrazolidine
IUPAC Name 1,2-diphenylpyrazolidine-3,5-dione
SMILES O=C1CC(=O)N(N1c2ccccc2)c3ccccc3
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C15H12N2O2/c18-14-11-15(19)17(13-9-5-2-6-10-13)16(14)12-7-3-1-4-8-12/h1-10H,11H2
Formula Weight 252.0899
Chemical Name or Material 1,2-Diphenyl-3,5-pyrazolidinedione