CAS RN 2652-77-9
CAS RN 2652-77-9
IUPAC Name:
1,2-diphenylpyrazolidine-3,5-dione
Synonyms:
1,2-diphenylpyrazolidine-3,5-dione
Molecular Weight (g/mol):
252.27
Molecular Formula:
C15H12N2O2
InChi Key:
XDPKQGKEOCYMQC-UHFFFAOYSA-N
SMILES:
O=C1CC(=O)N(N1C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
1,2-Diphenyl-3,5-pyrazolidinedione, 98%, Thermo Scientific™
CAS: 2652-77-9 Molecular Formula: C15H12N2O2 Molecular Weight (g/mol): 252.273 MDL Number: MFCD00179924 InChI Key: XDPKQGKEOCYMQC-UHFFFAOYSA-N Synonym: 1,2-diphenyl-pyrazolidine-3,5-dione,diphenyldiketopyrazolidine,1,2-diphenyl-3,5-pyrazolidinedione,difenildichetopirazolidina,1,2-diphenyl-3,5-dioxopyrazolidine,difenildichetopirazolidina italian,1,2-diphenyl-3,5-dioxopyrazolidin,3,5-pyrazolidinedione, 1,2-diphenyl,1,2-diphenyl-3,5-dioxopyrazolidin german,1,2-diphenyl-1,2-diazolidine-3,5-dione PubChem CID: 17561 IUPAC Name: 1,2-diphenylpyrazolidine-3,5-dione SMILES: C1C(=O)N(N(C1=O)C2=CC=CC=C2)C3=CC=CC=C3