Learn More
1,2-Cyclohexanedicarboxylic acid, bis(2-ethylhexyl) ester 100 μg/mL in Hexane, Dr. Ehrenstorfer
Product Code. 30309162
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309162 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30309162 Supplier Dr. Ehrenstorfer Supplier No. DREA11824550HE100
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,2-Cyclohexanedicarboxylic acid, bis(2-ethylhexyl) ester
CAS 84-71-9
Analyte or Component Names 1,2-Cyclohexanedicarboxylic acid, bis(2-ethylhexyl) ester
Shipping Condition +20°C
Molecular Formula C24 H44 O4
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 1,2-Cyclohexanedicarboxylic acid, bis(2-ethylhexyl) ester (6CI,7CI,8CI,9CI), Bis(2-ethylhexyl) cyclohexane-1,2-carboxylate, Bis(2-ethylhexyl) cyclohexane-1,2-dicarboxylate, Bis(2-ethylhexyl) hexahydrophthalate, Di(2-ethylhexyl) hexahydrophthalate, Diisooctyl 1,2-cyclohexanedicarboxylate, Dioctyl 1,2-cyclohexanedicarboxylate, Nan Ya DPEH
InChI Formula InChI=1S/C24H44O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h19-22H,5-18H2,1-4H3
SMILES CCCCC(CC)COC(=O)C1CCCCC1C(=O)OCC(CC)CCCC
IUPAC Name bis(2-ethylhexyl) cyclohexane-1,2-dicarboxylate
Solvent or Matrix Hexane
Molecular Weight (g/mol) 396.6
Quantity 1 mL
Formula Weight 396.324 g/mol