Learn More
1,2,5,6-Tetrabromocyclooctane, TRC
Product Code. 30136860
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
50 mg
Unit Size:
10mg
2.5mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136860 50 mg 50mg
30009294 2.5 mg 2.5mg
30006449 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136860 Supplier Toronto Research Chemicals Supplier No. TRCT29113050MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3194-57-8
Molecular Formula C8 H12 Br4
Molecular Weight (g/mol) 427.8
Synonym 1,2,5,6-Tetrabromocyclooctane, NSC 167079
IUPAC Name 1,2,5,6-tetrabromocyclooctane
SMILES BrC1CCC(Br)C(Br)CCC1Br
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C8H12Br4/c9-5-1-2-6(10)8(12)4-3-7(5)11/h5-8H,1-4H2
Formula Weight 423.7673
Chemical Name or Material 1,2,5,6-Tetrabromocyclooctane