CAS RN 3194-57-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,2,5,6-Tetrabromocyclooctane, TRC
CAS: 3194-57-8 Molecular Formula: C8 H12 Br4 Molecular Weight (g/mol): 427.8 Synonym: 1,2,5,6-Tetrabromocyclooctane,NSC 167079 IUPAC Name: 1,2,5,6-tetrabromocyclooctane SMILES: BrC1CCC(Br)C(Br)CCC1Br