missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1'-[Propane-2,2-diylbis-[sulfanediyl[(2S)-2-methyl-1-oxopropane-3,1-diyl]]]bis[(2S)-pyrrolidine-2-carboxylic] Acid, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
1,1'-[Propane-2,2-diylbis-[sulfanediyl[(2S)-2-methyl-1-oxopropane-3,1-diyl]]]bis[(2S)-pyrrolidine-2-carboxylic] Acid
Specifications
Specifications
| Chemical Name or Material | 1,1'-[Propane-2,2-diylbis-(sulfanediyl((2S)-2-methyl-1-oxopropane-3,1-diyl))]bis[(2S)-pyrrolidine-2-carboxylic] Acid |
| Analyte or Component Names | 1,1'-[Propane-2,2-diylbis-(sulfanediyl((2S)-2-methyl-1-oxopropane-3,1-diyl))]bis[(2S)-pyrrolidine-2-carboxylic] Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H34 N2 O6 S2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Captopril Imp. O (EP), 1,1'-[Propane-2,2-diylbis-[sulfanediyl[(2S)-2-methyl-1-oxopropane-3,1-diyl]]]bis[(2S)-pyrrolidine-2-carboxylic] Acid, Captopril Impurity O |
| InChI Formula | InChI=1S/C16H16O2/c1-12(18-2)14-9-6-10-15(11-14)16(17)13-7-4-3-5-8-13/h3-12H,1-2H7 |
| SMILES | C[C@H](CSC(C)(C)SC[C@@H](C)C(=O)N1CCC[C@H]1C(=O)O)C(=O)N2CCC[C@H]2C(=O)O |
| IUPAC Name | (2S)-1-[(2S)-3-[2-[(2S)-3-[(2S)-2-carboxypyrrolidin-1-yl]-2-methyl-3-oxopropyl]sulfanylpropan-2-ylsulfanyl]-2-methylpropanoyl]pyrrolidine-2-carboxylic acid |
| Molecular Weight (g/mol) | 474.63 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction