Learn More
1,1-Diphenyl-3-(N-piperidino)prop-1-ene Hydrochloride, Mikromol™
Product Code. 17974206
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17974206 25 mg 25mg
17940486 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17974206 Supplier Mikromol Supplier No. MM3493.010025
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,1-Diphenyl-3-(N-piperidino)prop-1-ene Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,1-Diphenyl-3-(N-piperidino)prop-1-ene Hydrochloride
CAS 30778-27-9
Analyte or Component Names 1,1-Diphenyl-3-(N-piperidino)prop-1-ene Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C20 H23 N . Cl H
Certifications/Compliance ISO 17025
Synonym 1,1-Diphenyl-3-(N-piperidino)prop-1-ene Hydrochloride, 1-(3,3-diphenyl-2-propen-1-yl)-Piperidine hydrochloride (1:1), Piperidine, 1-(3,3-diphenyl-2-propenyl)-, hydrochloride (9CI), Piperidine, 1-(3,3-diphenylallyl)-, hydrochloride (6CI,8CI), 1,1-Diphenyl-3-N-piperidino-prop-1-ene Hydrochloride, Pridinol Hydrochloride Imp. B as Hydrochloride
InChI Formula InChI=1S/C20H23N.ClH/c1-4-10-18(11-5-1)20(19-12-6-2-7-13-19)14-17-21-15-8-3-9-16-21;/h1-2,4-7,10-14H,3,8-9,15-17H2;1H
SMILES Cl.C(C=C(c1ccccc1)c2ccccc2)N3CCCCC3
IUPAC Name 1-(3,3-diphenylprop-2-enyl)piperidine;hydrochloride
Molecular Weight (g/mol) 313.86
Quantity 25 mg
Formula Weight 313.1597 g/mol