Learn More
1,1-Dichloro-1-(difluoromethoxy)-2,2,2-trifluoroethane, Mikromol™
Product Code. 30051144
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051144 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30051144 Supplier Mikromol Supplier No. MM0500.04
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,1-Dichloro-1-(difluoromethoxy)-2,2,2-trifluoroethane
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,1-Dichloro-1-(difluoromethoxy)-2,2,2-trifluoroethane
CAS 32778-07-7
Analyte or Component Names 1,1-Dichloro-1-(difluoromethoxy)-2,2,2-trifluoroethane
Shipping Condition Ice pack (-20°C)
Molecular Formula C3 H Cl2 F5 O
Certifications/Compliance ISO 9001
Synonym Isoflurane Imp. D (EP), 1,1-Dichloro-1-(difluoromethoxy)-2,2,2-trifluoroethane, Isoflurane Impurity D
InChI Formula InChI=1S/C3HCl2F5O/c4-2(5,3(8,9)10)11-1(6)7/h1H
SMILES FC(F)OC(Cl)(Cl)C(F)(F)F
IUPAC Name 1,1-dichloro-1-(difluoromethoxy)-2,2,2-trifluoroethane
Molecular Weight (g/mol) 218.94
Quantity 100 mg
Formula Weight 217.9325 g/mol