missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E. Coli J5 LPS Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB100-62551
This item is not returnable.
View return policy
Description
E. Coli J5 LPS Polyclonal specifically detects E. Coli J5 LPS in Bacteria samples. It is validated for ELISA.
Specifications
| E. Coli J5 LPS | |
| Polyclonal | |
| ELISA 1:100-1:2000 | |
| Escherichia coli | |
| Heat inactivated Escherichia coli J5 (Rc mutant; ATCC) | |
| 0.5 mL | |
| Primary | |
| Bacteria | |
| Antisera |
| ELISA | |
| Unconjugated | |
| Whole antisera with 0.09% Sodium Azide | |
| Sheep | |
| Unpurified | |
| RUO | |
| NB100-62551 recognizes the J5 mutant of E. coli 0:111 which lacks the enzyme uridine diphosphate glucose 4-epimerase and therefore produces an incomplete LPS, deficient in galactose and all the sugars distal to the central polymers. The 55 mutant has no O-specific chains and its endotoxin remains as the core LPS containing lipid A, N acetyl glucosamine, 2-keto-3-deoxyoctonate, heptose and glucose (a composition similar to that of the Rc strains of Salmonella). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction