missing translation for 'onlineSavingsMsg'
Learn More

E. Coli J5 LPS Antibody, Novus Biologicals™

Sheep Polyclonal Antibody

193.00 € - 501.00 €

Specifications

Antigen E. Coli J5 LPS
Dilution ELISA 1:100-1:2000
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18075116
View Documents
Novus Biologicals
NB100-62551-0.1ML
0.1 mL
193.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18708823
View Documents
Novus Biologicals
NB100-62551
0.5 mL
501.00 €
0.50mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

E. Coli J5 LPS Polyclonal specifically detects E. Coli J5 LPS in Bacteria samples. It is validated for ELISA.
Specifications

Specifications

E. Coli J5 LPS
ELISA
Unconjugated
Sheep
Bacteria
Escherichia coli
IgG
Unpurified
NB100-62551 recognizes the J5 mutant of E. coli 0:111 which lacks the enzyme uridine diphosphate glucose 4-epimerase and therefore produces an incomplete LPS, deficient in galactose and all the sugars distal to the central polymers. The 55 mutant has no O-specific chains and its endotoxin remains as the core LPS containing lipid A, N acetyl glucosamine, 2-keto-3-deoxyoctonate, heptose and glucose (a composition similar to that of the Rc strains of Salmonella).
ELISA 1:100-1:2000
Polyclonal
Antisera
RUO
Whole antisera with 0.09% Sodium Azide
Heat inactivated Escherichia coli J5 (Rc mutant; ATCC)
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.