accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NHSNH2CH3

CAS RN 97480-60-9

IUPAC Name: (3,5-dimethylphenyl)thiourea
Synonyms: 3,5-dimethylphenylthiourea
Molecular Weight (g/mol): 180.27
Molecular Formula: C9H12N2S
InChi Key: XOAYHDJRYDSPJZ-UHFFFAOYSA-N
SMILES: CC1=CC(NC(N)=S)=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results