accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 97-41-6

CH3OOH3CCH3H3CCH3

CAS RN 97-41-6

IUPAC Name: ethyl 2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropane-1-carboxylate
Synonyms: ethyl chrysanthemate
Molecular Weight (g/mol): 196.29
Molecular Formula: C12H20O2
InChi Key: VIMXTGUGWLAOFZ-UHFFFAOYNA-N
SMILES: CCOC(=O)C1C(C=C(C)C)C1(C)C
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)