Learn More
Ethyl Chrysanthemate, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | Ethyl Chrysanthemate |
| CAS | 97-41-6 |
| Molecular Formula | C12H20O2 |
| Quantity | 100 g |
| Synonym | 2,2-Dimethyl-3-(2-methyl-1-propen-1-yl)cyclopropanecarboxylic Acid Ethyl Ester, Chrysanthemummonocarboxylic Acid Ethyl Ester, 2,2-dimethyl-3-(2-methylpropenyl)cyclopropanecarboxylic Acid Ethyl Ester, Chrysanthemic Acid Ethyl Ester, Chrysanthemumic Acid Ethyl Ester, Ethyl 2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropanecarboxylate, Ethyl chrysanthemumate, NSC 10819 |
| InChI Formula | InChI=1S/C12H20O2/c1-6-14-11(13)10-9(7-8(2)3)12(10,4)5/h7,9-10H,6H2,1-5H3 |
| SMILES | CCOC(=O)C1C(C=C(C)C)C1(C)C |
| Molecular Weight (g/mol) | 196.29 |
| Formula Weight | 196.1463 |