accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 969-33-5

HClCH3N

CAS RN 969-33-5

IUPAC Name: hydrogen 1-methyl-4-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}piperidine chloride
Synonyms: hydrogen cyproheptadine chloride
Molecular Weight (g/mol): 323.86
Molecular Formula: C21H22ClN
InChi Key: ZPMVNZLARAEGHB-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12
Color 
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results