Learn More
Cyproheptadine hydrochloride, MedChemExpress
Product Code. 16488256
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
50mg
Product Code. 16488256
Brand: Medchem Express HYB0366A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Cyproheptadine hydrochloride is a potent and orally active 5-HT2A receptor antagonist, with antidepressant and antiserotonergic effects. Cyproheptadine hydrochloride has antiplatelet and thromboprotective activities. Cyproheptadine hydrochloride can be used for the research of thromboembolic disorders.
Cyproheptadine hydrochloride is a potent and orally active 5-HT2A receptor antagonist, with antidepressant and antiserotonergic effects. Cyproheptadine hydrochloride has antiplatelet and thromboprotective activities. Cyproheptadine hydrochloride can be used for the research of thromboembolic disorders.
Specifications
Chemical Name or Material Cyproheptadine hydrochloride
Physical Form Solid
CAS 969-33-5
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H22ClN
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 50 mg/mL (154.39 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.07 mM; Need ultrasonic)
SMILES CN1CC/C(CC1)=C2C3=CC=CC=C3C=CC4=CC=CC=C/24.Cl
Molecular Weight (g/mol) 323.86
Formula Weight 323.86
Percent Purity 99.98%
Grade Research