CAS RN 95407-69-5
IUPAC Name:
4-methoxy-2,2,6,6-tetramethylpiperidin-1-olate
Synonyms:
4-methoxy-2,2,6,6-tetramethylpiperidin-1-olate
Molecular Weight (g/mol):
186.28
Molecular Formula:
C10H20NO2
InChi Key:
BGXWUNHAMWRFOR-UHFFFAOYSA-N
SMILES:
COC1CC(C)(C)N([O-])C(C)(C)C1
1
–
1
of
1
results
1
4-Methoxy-TEMPO, free radical, 98+%, Thermo Scientific™
CAS: 95407-69-5 Molecular Formula: C10H20NO2 MDL Number: MFCD00270334 Synonym: 4-Methoxy-2,2,6,6-tetramethylpiperidinyloxy, free radical