accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3ONO

CAS RN 953408-85-0

IUPAC Name: 4-methyl-2-phenyl-1,3-oxazole-5-carbaldehyde
Synonyms: 4-methyl-2-phenyl-1,3-oxazole-5-carbaldehyde
Molecular Weight (g/mol): 187.2
Molecular Formula: C11H9NO2
InChi Key: NIYJTYPVRBYCEZ-UHFFFAOYSA-N
SMILES: CC1=C(OC(=N1)C1=CC=CC=C1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results