accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 950858-06-7

FHOFI

CAS RN 950858-06-7

IUPAC Name: 2,6-difluoro-4-iodophenol
Synonyms: 2,6-difluoro-4-iodophenol
Molecular Weight (g/mol): 255.99
Molecular Formula: C6H3F2IO
InChi Key: LHUADYKKTXZZMQ-UHFFFAOYSA-N
SMILES: OC1=C(F)C=C(I)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results