accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 937796-14-0

CH3HNNNNO

CAS RN 937796-14-0

IUPAC Name: methyl({[2-(morpholin-4-yl)pyrimidin-5-yl]methyl})amine
Synonyms: methyl({[2-(morpholin-4-yl)pyrimidin-5-yl]methyl})amine
Molecular Weight (g/mol): 208.27
Molecular Formula: C10H16N4O
InChi Key: PFYPMSRZQMFCJO-UHFFFAOYSA-N
SMILES: CNCC1=CN=C(N=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results