CAS RN 928659-70-5
CAS RN 928659-70-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Eliglustat hemitartrate, MedChemExpress
MedChemExpress Eliglustat hemitartrate is an specific, potent and orally active glucocerebroside synthase inhibitor with an IC50 of 24 nM.