Learn More
Eliglustat hemitartrate, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Description
Eliglustat hemitartrate is an specific, potent and orally active glucocerebroside synthase inhibitor with an IC50 of 24 nM.
Specifications
Specifications
| Chemical Name or Material | Eliglustat hemitartrate |
| CAS | 928659-70-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C50H78N4O14 |
| Quantity | 10 mg |
| Synonym | Genz-112638 Eliglustat tartrate |
| Solubility Information | DMSO : 110 mg/mL (114.68 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (52.13 mM) |
| SMILES | CCCCCCCC(N[C@H](CN1CCCC1)[C@@H](C2=CC=C(OCCO3)C3=C2)O)=O.CCCCCCCC(N[C@H](CN4CCCC4)[C@@H](C5=CC=C(OCCO6)C6=C5)O)=O.O=C(O)[C@H](O)[C@@H](O)C(O)=O |
| Molecular Weight (g/mol) | 959.17 |
| Show More |