CAS RN 92136-39-5
CAS RN 92136-39-5
IUPAC Name:
tert-butyl N-(prop-2-yn-1-yl)carbamate
Synonyms:
tert-butyl N-(prop-2-yn-1-yl)carbamate
Molecular Weight (g/mol):
155.2
Molecular Formula:
C8H13NO2
InChi Key:
DSPYCWLYGXGJNJ-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NCC#C
1
–
1
of
1
results
1
N-Boc-propargylamine, 97%
CAS: 92136-39-5 Molecular Formula: C8H13NO2 Molecular Weight (g/mol): 155.197 MDL Number: MFCD07367245 InChI Key: DSPYCWLYGXGJNJ-UHFFFAOYSA-N Synonym: n-boc-propargylamine,n-tert-butoxycarbonyl propargylamine,n-boc-prop-2-ynylamine,tert-butyl prop-2-yn-1-ylcarbamate,n-boc-propargyl amine,tert-butyl n-prop-2-yn-1-yl carbamate,tert-butyl prop-2-ynylcarbamate,n-boc-2-propyn-1-amine,n-boc propargyl amine,prop-2-ynyl-carbamic acid tert-butyl ester PubChem CID: 11073648 IUPAC Name: tert-butyl N-prop-2-ynylcarbamate SMILES: CC(C)(C)OC(=O)NCC#C