Learn More
N-Boc-propargylamine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | N-Boc-propargylamine |
| CAS | 92136-39-5 |
| Molecular Formula | C8H13NO2 |
| Quantity | 5 g |
| Synonym | N-2-Propyn-1-yl-carbamic Acid 1,1-Dimethylethyl Ester, 2-Propynyl-carbamic Acid 1,1-Dimethylethyl Ester, 1,1-Dimethylethyl 2-Propyn-1-ylcarbamate, 2-Propynylcarbamic Acid tert-Butyl Ester, 3-(tert-Butoxycarbonylamino)prop-1-yne, 3-[(tert-Butyloxycarbonyl)amino]-1-propyne, N-(Prop-2-ynyl)carbamic acid tert-butyl ester, N-(tert-Butoxycarbonyl)propargylamine, N-Boc-propargylamine, N-tert-Butoxycarbonyl-2-propyn-1-amine, tert-Butyl (2-Propyn-1-yl)carbamate, tert-Butyl 2-Propynylcarbamate, tert-Butyl N-Propargylcarbamate, tert-Butyl Propargylcarbamate |
| InChI Formula | InChI=1S/C8H13NO2/c1-5-6-9-7(10)11-8(2,3)4/h1H,6H2,2-4H3,(H,9,10) |
| SMILES | CC(C)(C)OC(=O)NCC#C |
| IUPAC Name | tert-butyl N-prop-2-ynylcarbamate |
| Molecular Weight (g/mol) | 155.2 |
| Formula Weight | 155.0946 |