accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 92-91-1

H3CO

CAS RN 92-91-1

IUPAC Name: 1-{[1,1'-biphenyl]-4-yl}ethan-1-one
Synonyms: 4-acetylbiphenyl
Molecular Weight (g/mol): 196.25
Molecular Formula: C14H12O
InChi Key: QCZZSANNLWPGEA-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results