CAS RN 917837-54-8
IUPAC Name:
N-(4-chlorophenyl)-3-[(4-chlorophenyl)amino]-2-(3-methyl-1,2-oxazol-5-yl)prop-2-enamide
Synonyms:
N-(4-chlorophenyl)-3-[(4-chlorophenyl)amino]-2-(3-methyl-1,2-oxazol-5-yl)prop-2-enamide
Molecular Weight (g/mol):
388.25
Molecular Formula:
C19H15Cl2N3O2
InChi Key:
VMAKIACTLSBBIY-UHFFFAOYSA-N
SMILES:
CC1=NOC(=C1)C(=CNC1=CC=C(Cl)C=C1)C(=O)NC1=CC=C(Cl)C=C1
1
–
2
of
2
results
1
Tocris Bioscience™ CCMI
CAS: 917837-54-8 Molecular Formula: C19H15Cl2N3O2
2
CCMI, MedChemExpress
MedChemExpress CCMI (AVL-3288) is a potent and selective α7 nAChR-positive allosteric modulator, does not bind to or activate α7 nAChRs via the orthosteric site, and causes significant positive modulation of agonist-induced currents at α7 nAChRs. CCMI has potential in CNS diseases with cognitive dysfunction.