Learn More
CCMI, MedChemExpress
Product Code. 16467727
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467727 5 mg 5mg
16454998 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16467727 Supplier Medchem Express Supplier No. HY12150
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
MedChemExpress CCMI (AVL-3288) is a potent and selective α7 nAChR-positive allosteric modulator, does not bind to or activate α7 nAChRs via the orthosteric site, and causes significant positive modulation of agonist-induced currents at α7 nAChRs. CCMI has potential in CNS diseases with cognitive dysfunction.
CCMI (AVL-3288) is a potent and selective α7 nAChR-positive allosteric modulator, does not bind to or activate α7 nAChRs via the orthosteric site, and causes significant positive modulation of agonist-induced currents at α7 nAChRs. CCMI has potential in CNS diseases with cognitive dysfunction.
Specifications
Chemical Name or Material CCMI
Physical Form Solid
CAS 917837-54-8
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C19H15Cl2N3O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym AVL-3288 UCI-4083
Solubility Information DMSO : 100 mg/mL (257.57 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(Cl)C=C1)/C(C2=CC(C)=NO2)=C/NC3=CC=C(Cl)C=C3
Molecular Weight (g/mol) 388.25
Formula Weight 388.25
Percent Purity 99.32%
Grade Research