accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 916420-54-7

CH3OFFFOH

CAS RN 916420-54-7

IUPAC Name: [3-methyl-5-(trifluoromethoxy)phenyl]methanol
Synonyms: [3-methyl-5-(trifluoromethoxy)phenyl]methanol
Molecular Weight (g/mol): 206.16
Molecular Formula: C9H9F3O2
InChi Key: JELFBZMXRZYDFP-UHFFFAOYSA-N
SMILES: CC1=CC(CO)=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results