accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 913835-95-7

BSOONOH3CHOOH

CAS RN 913835-95-7

IUPAC Name: (4-{benzyl[(4-methoxyphenyl)methyl]sulfamoyl}phenyl)boronic acid
Synonyms: 4-{benzyl[(4-methoxyphenyl)methyl]sulfamoyl}phenylboronic acid
Molecular Weight (g/mol): 411.28
Molecular Formula: C21H22BNO5S
InChi Key: KVYLYENGQVBOPD-UHFFFAOYSA-N
SMILES: COC1=CC=C(CN(CC2=CC=CC=C2)S(=O)(=O)C2=CC=C(C=C2)B(O)O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results