accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 913835-74-2

BFOHHOOH

CAS RN 913835-74-2

IUPAC Name: (4-fluoro-3-hydroxyphenyl)boronic acid
Synonyms: 4-fluoro-3-hydroxyphenylboronic acid
Molecular Weight (g/mol): 155.92
Molecular Formula: C6H6BFO3
InChi Key: GNCXNRGBOBWFSO-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results