accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 91053-39-3

OSiO

CAS RN 91053-39-3

IUPAC Name: silanedione
Synonyms: silica 二氧化硅 siliziumdioxid solum diatomeae quartz (SiO2)n kieselsaeureanhydrid SiO2 silica gel silica dimethyl silylate
Molecular Weight (g/mol): 60.08
Molecular Formula: O2Si
InChi Key: VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES: O=[Si]=O
Quantity 
  • (3)
  • (1)
  • (1)
Color 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)