accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NH3CONNNHHCl

CAS RN 902836-08-2

IUPAC Name: N,N-dimethyl-2-(piperazin-1-yl)pyridine-3-carboxamide hydrochloride
Synonyms: N,N-dimethyl-2-(piperazin-1-yl)pyridine-3-carboxamide hydrochloride
Molecular Weight (g/mol): 270.76
Molecular Formula: C12H19ClN4O
InChi Key: YRPLJKUUYNCGQS-UHFFFAOYSA-N
SMILES: Cl.CN(C)C(=O)C1=C(N=CC=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results