accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SiOHMen

CAS RN 9004-73-3

IUPAC Name: 2-methyl-1,2-oxasilinan-2-yl
Synonyms: methicone polymethylhydrosiloxane
Molecular Weight (g/mol): 115.23
Molecular Formula: C5H11OSi
InChi Key: TXYXVOOPRILIHR-UHFFFAOYSA-N
SMILES: C[Si]1CCCCO1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results