accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 90-26-6

CH3NH2O

CAS RN 90-26-6

IUPAC Name: 2-phenylbutanamide
Synonyms: benzeneacetamide, α-ethyl-
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: UNFGQCCHVMMMRF-UHFFFAOYNA-N
SMILES: CCC(C(N)=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results