accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 89551-24-6

H2N

CAS RN 89551-24-6

IUPAC Name: 1-phenylmethanamine
Synonyms: benzylamine sumine 2005 sumine 2006 moringine ω-aminotoluene
Molecular Weight (g/mol): 107.16
Molecular Formula: C7H9N
InChi Key: WGQKYBSKWIADBV-UHFFFAOYSA-N
SMILES: NCC1=CC=CC=C1
Quantity 
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (3)
  • (3)