accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 89488-25-5

BClOHHOOH

CAS RN 89488-25-5

IUPAC Name: (5-chloro-2-hydroxyphenyl)boronic acid
Synonyms: 5-chloro-2-hydroxyphenylboronic acid
Molecular Weight (g/mol): 172.37
Molecular Formula: C6H6BClO3
InChi Key: GGZASRFCRAZNPO-UHFFFAOYSA-N
SMILES: OB(O)C1=C(O)C=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results