accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 88878-50-6

H3COOF

CAS RN 88878-50-6

IUPAC Name: 4-fluorophenyl 4'-pentyl-[1,1'-bi(cyclohexane)]-4-carboxylate
Synonyms: 4-fluorophenyl 4'-pentyl-[1,1'-bi(cyclohexane)]-4-carboxylate
Molecular Weight (g/mol): 374.54
Molecular Formula: C24H35FO2
InChi Key: ZVCPJOLFAYLVER-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(CC1)C1CCC(CC1)C(=O)OC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results