accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 886502-76-7

CH3FFFO

CAS RN 886502-76-7

IUPAC Name: 5-methyl-2-(trifluoromethyl)benzaldehyde
Synonyms: 5-methyl-2-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 188.15
Molecular Formula: C9H7F3O
InChi Key: ZXQHVZWHSXZEQV-UHFFFAOYSA-N
SMILES: CC1=CC(C=O)=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results