accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 886501-03-7

CH3FNH2Cl

CAS RN 886501-03-7

IUPAC Name: 2-chloro-6-fluoro-3-methylaniline
Synonyms: 2-chloro-6-fluoro-3-methylaniline
Molecular Weight (g/mol): 159.59
Molecular Formula: C7H7ClFN
InChi Key: ZEDOBEHSBNTVKR-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C(N)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results