accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 886498-17-5

OFFFOH2NF

CAS RN 886498-17-5

IUPAC Name: 2-fluoro-5-(trifluoromethoxy)benzamide
Synonyms: 2-fluoro-5-(trifluoromethoxy)benzamide
Molecular Weight (g/mol): 223.13
Molecular Formula: C8H5F4NO2
InChi Key: NYHOMJMCMKWNKT-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(F)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results