accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 87594-59-0

OHONOOOH2N

CAS RN 87594-59-0

IUPAC Name: 4-carbamoyl-3-nitrobenzoic acid
Synonyms: 4-carbamoyl-3-nitrobenzoic acid
Molecular Weight (g/mol): 210.15
Molecular Formula: C8H6N2O5
InChi Key: LKPGLAHLFCDSNK-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(C=C(C=C1)C(O)=O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results