accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CCH3OONNClNOO

CAS RN 874814-97-8

IUPAC Name: tert-butyl 4-(4-chloro-2-nitrophenyl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(4-chloro-2-nitrophenyl)piperazine-1-carboxylate
Molecular Weight (g/mol): 341.79
Molecular Formula: C15H20ClN3O4
InChi Key: OJIDLANQYRRUCV-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=C(C=C(Cl)C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results