accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BSOONHOOH

CAS RN 871329-68-9

IUPAC Name: [4-(azetidine-1-sulfonyl)phenyl]boronic acid
Synonyms: 4-(azetidine-1-sulfonyl)phenylboronic acid
Molecular Weight (g/mol): 241.07
Molecular Formula: C9H12BNO4S
InChi Key: IOGLONDWRZETTA-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)S(=O)(=O)N1CCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results